Anti-NUP160 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
USBI039216-BIOTINEA 852 EUR
USBI039216-BIOTIN
Anti-NUP160 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: NUP160 (nucleoporin 160kDa)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR