Anti-NXF1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: US Biological
039228-AP
USBI039228-APEA 852 EUR
USBI039228-AP
Anti-NXF1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: NXF1 (nuclear RNA export factor 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR