Anti-ONECUT2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: US Biological
039283-PE
USBI039283-PEEA 852 EUR
USBI039283-PE
Anti-ONECUT2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: ONECUT2 (one cut homeobox 2)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR