Anti-OR10A4 Rabbit Polyclonal Antibody

Supplier: US Biological
039296
USBI039296EA 587 EUR
USBI039296
Anti-OR10A4 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: OR10A4 (olfactory receptor, family 10, subfamily A, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR