Anti-OR10J3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
039309-APC
USBI039309-APCEA 852 EUR
USBI039309-APC
Anti-OR10J3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: OR10J3 (olfactory receptor, family 10, subfamily J, member 3)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR