Anti-OR14C36 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: US Biological
039326-AP
USBI039326-APEA 852 EUR
USBI039326-AP
Anti-OR14C36 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: OR14C36 (olfactory receptor, family 14, subfamily C, member 36)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR