Anti-OR2A12 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: US Biological
039342-FITC
USBI039342-FITCEA 852 EUR
USBI039342-FITC
Anti-OR2A12 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: OR2A12 (olfactory receptor, family 2, subfamily A, member 12)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR