Anti-OR2A4 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
039343-BIOTIN
USBI039343-BIOTINEA 852 EUR
USBI039343-BIOTIN
Anti-OR2A4 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: OR2A4 (olfactory receptor, family 2, subfamily A, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR