Anti-OR2M3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: US Biological
039362-PE
USBI039362-PEEA 852 EUR
USBI039362-PE
Anti-OR2M3 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: OR2M3 (olfactory receptor, family 2, subfamily M, member 3)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR