Anti-OR2T3 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
039369-BIOTIN
USBI039369-BIOTINEA 852 EUR
USBI039369-BIOTIN
Anti-OR2T3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50

Type: Primary
Antigen: OR2T3 (olfactory receptor, family 2, subfamily T, member 3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR