Anti-OR2W3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
039377-APC
USBI039377-APCEA 852 EUR
USBI039377-APC
Anti-OR2W3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: OR2W3 (olfactory receptor, family 2, subfamily W, member 3)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR