Anti-OR4C13 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
039386-BIOTIN
USBI039386-BIOTINEA 852 EUR
USBI039386-BIOTIN
Anti-OR4C13 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: OR4C13 (olfactory receptor, family 4, subfamily C, member 13)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR