Anti-OR4F15 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
039392-HRP
USBI039392-HRPEA 852 EUR
USBI039392-HRP
Anti-OR4F15 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: OR4F15 (olfactory receptor, family 4, subfamily F, member 15)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR