Anti-OR4K5 Rabbit Polyclonal Antibody

Supplier: US Biological
039403
USBI039403EA 587 EUR
USBI039403
Anti-OR4K5 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: OR4K5 (olfactory receptor, family 4, subfamily K, member 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR