Anti-OR51T1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: US Biological
039426-AP
USBI039426-APEA 852 EUR
USBI039426-AP
Anti-OR51T1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: OR51T1 (olfactory receptor, family 51, subfamily T, member 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR