Anti-OR5AK2 Rabbit Polyclonal Antibody

Supplier: US Biological
039447
USBI039447EA 587 EUR
USBI039447
Anti-OR5AK2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: OR5AK2 (olfactory receptor, family 5, subfamily AK, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR