Anti-P-Glycoprotein Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
039557-HRP
USBI039557-HRPEA 852 EUR
USBI039557-HRP
Anti-P-Glycoprotein Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: P-Glycoprotein
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR