Anti-CDKN1A Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: US Biological
039564-PE
USBI039564-PEEA 852 EUR
USBI039564-PE
Anti-CDKN1A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: FLISA: 1:1000; Dot blot 1:500
Do not freeze!

Type: Primary
Antigen: CDKN1A (cyclin-dependent kinase inhibitor 1A (p21, Cip1))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR