Anti-CDKN1B Rabbit Polyclonal Antibody

Supplier: US Biological
USBI039570EA 587 EUR
USBI039570
Anti-CDKN1B Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Dot blot 1:500

Type: Primary
Antigen: CDKN1B (cyclin-dependent kinase inhibitor 1B (p27, Kip1))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR