Anti-P2RX6 Rabbit Polyclonal Antibody

Supplier: US Biological
039580
USBI039580EA 587 EUR
USBI039580
Anti-P2RX6 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: P2RX6 (purinergic receptor P2X, ligand-gated ion channel, 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR