Anti-P2RX7 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: US Biological
USBI039581-APEA 852 EUR
USBI039581-AP
Anti-P2RX7 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: P2RX7 (purinergic receptor P2X, ligand-gated ion channel, 7)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR