Anti-PABPC4L Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
039601-APC
USBI039601-APCEA 852 EUR
USBI039601-APC
Anti-PABPC4L Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: PABPC4L (poly(A) binding protein, cytoplasmic 4-like)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR