Anti-PABPN1L Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: US Biological
039603-PE
USBI039603-PEEA 852 EUR
USBI039603-PE
Anti-PABPN1L Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: PABPN1L (poly(A) binding protein, nuclear 1-like (cytoplasmic))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR