Anti-PAFAH1B2 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI039610EA 587 EUR
USBI039610
Anti-PAFAH1B2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Dot Blot 1:500

Type: Primary
Antigen: PAFAH1B2 (platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR