Anti-PAFAH2 Rabbit Polyclonal Antibody

Supplier: US Biological
039613
USBI039613EA 587 EUR
USBI039613
Anti-PAFAH2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: PAFAH2 (platelet-activating factor acetylhydrolase 2, 40kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR