Anti-PAK4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: US Biological
039627-PE
USBI039627-PEEA 852 EUR
USBI039627-PE
Anti-PAK4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: PAK4 (p21 protein (Cdc42/Rac)-activated kinase 4)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR