Anti-PARP1 Rabbit Polyclonal Antibody

Supplier: US Biological
039665
USBI039665EA 587 EUR
USBI039665
Anti-PARP1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: PARP1 (poly (ADP-ribose) polymerase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR