Anti-PARP1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: US Biological
039671-AP
USBI039671-APEA 852 EUR
USBI039671-AP
Anti-PARP1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Dot blot 1:500
Do not freeze!

Type: Primary
Antigen: PARP1 (poly (ADP-ribose) polymerase 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR