Anti-PAX5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
039702-APC
USBI039702-APCEA 852 EUR
USBI039702-APC
Anti-PAX5 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: PAX5 (paired box 5)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR