Anti-PAX5 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
039702-BIOTIN
USBI039702-BIOTINEA 852 EUR
USBI039702-BIOTIN
Anti-PAX5 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50; Immunochemistry: 1:10-50

Type: Primary
Antigen: PAX5 (paired box 5)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR