Anti-PAX7 Rabbit Polyclonal Antibody

Supplier: US Biological
039707
USBI039707EA 587 EUR
USBI039707
Anti-PAX7 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: PAX7 (paired box 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR