Anti-PBX3 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
039712-BIOTIN
USBI039712-BIOTINEA 852 EUR
USBI039712-BIOTIN
Anti-PBX3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: PBX3 (pre-B-cell leukemia homeobox 3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR