Anti-KAT2B Rabbit Polyclonal Antibody

Supplier: US Biological
039716
USBI039716EA 587 EUR
USBI039716
Anti-KAT2B Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100

Type: Primary
Antigen: KAT2B (K(lysine) acetyltransferase 2B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR