Anti-PCDHGC5 Rabbit Polyclonal Antibody

Supplier: US Biological
039761
USBI039761EA 587 EUR
USBI039761
Anti-PCDHGC5 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: PCDHGC5 (protocadherin gamma subfamily C, 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR