Anti-PDE3B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
039801-HRP
USBI039801-HRPEA 852 EUR
USBI039801-HRP
Anti-PDE3B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50

Type: Primary
Antigen: PDE3B (phosphodiesterase 3B, cGMP-inhibited)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR