Anti-PDE6A Rabbit Polyclonal Antibody

Supplier: US Biological
039805
USBI039805EA 587 EUR
USBI039805
Anti-PDE6A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: PDE6A (phosphodiesterase 6A, cGMP-specific, rod, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR