Anti-PDK2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: United States Biological
USBI039842-HRPEA 852 EUR
USBI039842-HRP
Anti-PDK2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: PDK2 (pyruvate dehydrogenase kinase, isozyme 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR