Anti-WWC1 Rabbit Polyclonal Antibody (Biotin)

Supplier: United States Biological
USBI043903-BIOTINEA 852 EUR
USBI043903-BIOTIN
Anti-WWC1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: WWC1 (WW and C2 domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR