Anti-SOCS4 Mouse Polyclonal Antibody

Supplier: United States Biological
USBI133685EA 522 EUR
USBI133685
Anti-SOCS4 Mouse Polyclonal Antibody
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: SOCS4 (suppressor of cytokine signaling 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR