Anti-SOD2 Mouse Polyclonal Antibody

Supplier: United States Biological
USBI133695EA 522 EUR
USBI133695
Anti-SOD2 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SOD2 (superoxide dismutase 2, mitochondrial)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR