Anti-SOHLH2 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI133698EA 567 EUR
USBI133698
Anti-SOHLH2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SOHLH2 (spermatogenesis and oogenesis specific basic helix-loop-helix 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR