Anti-SOX30 Mouse Polyclonal Antibody

Supplier: United States Biological
USBI133722EA 522 EUR
USBI133722
Anti-SOX30 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SOX30 (SRY (sex determining region Y)-box 30)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR