Anti-SOX6 Mouse Polyclonal Antibody

Supplier: United States Biological
USBI133727EA 522 EUR
USBI133727
Anti-SOX6 Mouse Polyclonal Antibody
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: SOX6 (SRY (sex determining region Y)-box 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR