Anti-SOX9 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI133729EA 561 EUR
USBI133729
Anti-SOX9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SOX9 (SRY (sex determining region Y)-box 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR