Anti-SUN5 Mouse Polyclonal Antibody

Supplier: United States Biological
USBI133741EA 522 EUR
USBI133741
Anti-SUN5 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SUN5 (Sad1 and UNC84 domain containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR