Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPAG8 Mouse Monoclonal Antibody [clone: 2F12]

Supplier: US Biological
Type: Primary
Antigen: SPAG8 (sperm associated antigen 8)
Clonality: monoclonal
Clone: 2F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG USBI133746
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol SPAG8
Antigen name sperm associated antigen 8
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 2F12
Host Mouse
Isotype IgG2a kappa
Western blot Yes
ELISA Yes
Purification purified by Protein A chromatography
Sequence MNGSTEGSRSRSRSLDIQPSSEGLGPTSEPFPSSDDSPRSALAAATAAAAAAASAAAATAAFTTAKAAALSTKTPAPCSEFMEPSSDPSLLGEPCAGPGFTHNIAHGSLGFEPVYVSCIAQDTCTTTDHSSNPGPVPGSSSGPVLGSSSGAGHGSGSGSGPGCGSVPGSGSGPGPGSGPGSGPGHGSGSHPGPASGPGPDTGPDSELSPCIPPGFRNLVADRVLNYTSWSQHCPWEPQKQPPWEFLQVLEPGA
Form liquid
Storage buffer PBS, pH 7,2.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty