Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SPG11 Mouse Polyclonal Antibody

Supplier: US Biological
Type: Primary
Antigen: SPG11 (spastic paraplegia 11 (autosomal recessive))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human



 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µg 50 µG USBI133760
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol SPG11
Antigen name spastic paraplegia 11 (autosomal recessive)
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Mouse
Isotype IgG
Western blot Yes
Purification purified by Protein A chromatography
Sequence MLRKILASQQPDRCKRAQAFISTQGLKPDTVAELVAEEVTRELLTSSQGTGHKQMFNPTEESQTFLQLTTLCQDRTLVGMKLLDKISSVPHGELSCTTELLILAHHCFTLTCHMEGIIRVLQAAHMLTDNHLAPSEEYGLVVRLLTGIGRYNEMTYIFDLLHKKHYFEVLMRKKLDPSGTLKTALLDYIKRCRPGDSEKHNMIALCFSMCREIGENHEAAARIQLKLIESQPWEDSLKDGHQLKQLLLKALTLMLDAAESYAKDSCVRQAQHCQRLTKLITLQIHFLNTGQNTMLINLGRHKLMDCILALPRFYQASIVAEAYDFVPDWAEILYQQVILKGDFNYLEEFKQQRLLKSSIFEEISKKYKQHQPTDMVMENLKKLLTYCEDVYLYYKLAYEHKFYEIVNVLLKDPQTGCCLKDMLAG
Form liquid
Storage buffer PBS, pH 7,2.
Storage temperature Cool
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping