Anti-SPHK1 Mouse Polyclonal Antibody

Supplier: United States Biological
USBI133765EA 522 EUR
USBI133765
Anti-SPHK1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SPHK1 (sphingosine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR