Anti-SPINK1 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI133773EA 561 EUR
USBI133773
Anti-SPINK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SPINK1 (serine peptidase inhibitor, Kazal type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR