Anti-SPO11 Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI133785EA 561 EUR
USBI133785
Anti-SPO11 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SPO11 (SPO11 meiotic protein covalently bound to DSB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR